Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dsRNA Mouse anti-Broad Species Reactivity, Clone: K1, Novus Biologicals™

Mouse Monoclonal Antibody
$771.00 - $1207.00
Specifications
Antigen | dsRNA |
---|---|
Clone | K1 |
Concentration | LYOPH |
Dilution | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Immunoaffinity Purification |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification |
Description
dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity PurificationSpecifications
dsRNA | |
LYOPH | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoblot, Affinity Purification | |
Unconjugated | |
Mouse | |
Multi-species | |
Double-stranded RNA | |
IgG2a κ | |
Protein A purified | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
K1 | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting, Immunoaffinity Purification | |
Monoclonal | |
Purified | |
RUO | |
Lyophilised from PBS | |
Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone. | |
Primary | |
The lyophilised sample should be reconstituted with 200 ul (500 ul for 500 ug size) sterile distilled water. The mAb will then be in PBS without any stabilisers or preservatives at a concentration of 1 mg/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title