Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsRNA Mouse anti-Broad Species Reactivity, Clone: K2, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$788.50 - $1240.00

Specifications

Antigen dsRNA
Clone K2
Dilution ELISA, Immunofluorescence
Applications ELISA, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121501
SDP
View Documents
Novus Biologicals
NBP3113985ML
5 mL
Each for $788.50
Add to Cart
 
NB121500
SDP
View Documents
Novus Biologicals
NBP31139810ML
10 mL
Each for $1,240.00
Add to Cart
 
Description

Description

dsRNA Monoclonal antibody specifically detects dsRNA in Broad Species Reactivity samples. It is validated for ELISA, Immunofluorescence
Specifications

Specifications

dsRNA
ELISA, Immunofluorescence
Monoclonal
Purified
RUO
Culture supernatant (RPMI, 5% fetal calf serum)
Female DBA/2 mice were injected intraperitonially with a mixture of 50 ug L-dsRNA and 75 ug methylated bovine serum albumin, emulsified in complete Freund's adjuvant. After several boosts spleen cells were fused with Sp2/0-Agl4 myeloma cells to generate the hybridoma clone.
Primary
Store at -20°C. Avoid freeze-thaw cycles.
K2
ELISA, Immunofluorescence
Unconjugated
Mouse
Multi-species
Double-stranded RNA
IgM κ
Unpurified
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit