Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSTN Mouse anti-Canine, Human, Clone: OTI1D8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502654

Catalog No. 50-167-3692


Add to cart

Description

Description

The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

DSTN
Monoclonal
0.91 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P60981
ACTDP, ADF, HEL32, bA462D18.2
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11034, 477147
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D8
Unconjugated
DSTN
Liquid
DSTN
Full length human recombit protein of human DSTN produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit