Learn More
Andwin Scientific Direct Thermal Cryo-Tags, 2.0X 0.75inch Orange
SDP
Supplier:  Andwin Scientific DTCR7000O
Encompass_Preferred
Direct Thermal Cryo-Tags are formatted to easily work with the Dymo Labelwriter, but also with most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both. Temperatures From 70 to -196degreeC.
Catalog No. NC1659361