Learn More
Diversified Biotech Inc DT TAG / SPOT COMBO WHITE
Supplier:  Diversified Biotech Inc DTCR6200
Encompass
DT Cryo-Tags 1.28" x 0.50" & 3/8" spotIdeal Use: 2.0ml tubes Colors: White, Blue, Green, Orange, Red, YellowU of M: 1,000/roll (500 tag and spot sets)Temperature Range: 70°C to -196°CDirect Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®. Quickly label your samples with sharp text, clear barcodes or a combination of both.
Catalog No. NC1864829
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.