Learn More
Dtk/TYRO3 Mouse anti-Human, Alexa Fluor 488, Clone: 96201, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB859G100UG
Catalog No. FAB859G100
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Dtk/TYRO3
96201
Flow Cytometry 0.25-1μg/106 cells
Q06418
Brt, BYK, Dtk, EC 2.7.10, EC 2.7.10.1, FLJ16467, RSE, Sky, Tif, TYRO3 protein tyrosine kinase, Tyrosine-protein kinase byk, Tyrosine-protein kinase DTK, tyrosine-protein kinase receptor TYRO3, Tyrosine-protein kinase RSE, Tyrosine-protein kinase SKY
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Tyro3/Dtk Ala41-Ser428 (predicted) Accession # Q06418
100μg
Primary
7301
Reconstitution default not known
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide.
Supernatant
Mouse
IgG1
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Tyro3/Dtk in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity or interferance with recombinant mouse Tyro3/Dtk, recombinant human (rh) Axl, or rhMer is observed.
Human