Learn More
Abnova™ DTNA 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00001837T03
Catalog No. 89-942-493
Add to cart
Specifications
Specifications
Western Blot
293T
BC005300.1
pCMV-DTNA full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DTNA 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 40.92kDa
Transient overexpression cell lysate was tested with anti-DTNA antibody (Mfr. No. H00001837-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)