Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTNA Mouse anti-Human, Clone: OTI1A2, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA502207
Description
The protein encoded by this gene belongs to the dystrobrevin subfamily of the dystrophin family. This protein is a component of the dystrophin-associated protein complex (DPC), which consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and alpha- and beta-dystrobrevin. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Mutations in this gene are associated with left ventricular noncompaction with congenital heart defects. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
DTNA | |
Monoclonal | |
0.66 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y4J8 | |
D18S892E, DRP3, DTN, DTN-A, LVNC1 | |
Mouse | |
IgG2a | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
1837 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1A2 | |
Unconjugated | |
DTNA | |
Liquid | |
DTNA | |
Full length human recombit protein of human DTNA produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title