Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTNB Mouse anti-Human, Mouse, Rat, Clone: OTI3H8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504332

Catalog No. 50-167-5050


Add to cart

Description

Description

This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications

Specifications

DTNB
Monoclonal
0.98 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60941, O70585, P84060
dystrobrevin beta
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13528, 1838, 362715
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI3H8
Unconjugated
DTNB
Liquid
DTNB
Full length human recombit protein of human DTNB produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit