Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DTNBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | DTNBP1 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5110982
|
Invitrogen™
PA5110982 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5110983
|
Invitrogen™
PA5110983 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: LNSGGEENTV LSPALGPESS TCQNEITLQV PNPSELRAKP PSSSSTCTDS ATRDISEGGE SPVVQSDE.
Dtnbp1 may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations are associated with Hermansky-Pudlak syndrome type 7. This protein may also be associated with schizophrenia.Specifications
DTNBP1 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96EV8 | |
84062 | |
Recombinant Protein corresonding to Human DTNBP1. Recombinant protein control fragment (Product #RP-94606). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
DTNBP1 | |
5430437B18Rik; AW048963; biogenesis of lysosomal organelles complex-1, subunit 8; biogenesis of lysosome-related organelles complex 1 subunit 8; BLOC-1 subunit 8; Bloc1s8; DBND; distrobrevin binding protein 1; DKFZp564K192; DTBP1; Dtnbp1; Dysbindin; dysbindin-1; dystrobrevin binding protein 1; dystrobrevin-binding protein 1; FLJ30031; Hermansky-Pudlak syndrome 7 protein; hermansky-Pudlak syndrome 7 protein homolog; HPS7; HPS7 protein; HPS7 protein homolog; Hps7-like protein; MGC20210; My031; OTTHUMP00000221462; OTTHUMP00000221485; RP1-147M19.1; sandy; sdy | |
DTNBP1 | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title