Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTX1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550650

Catalog No. PA550650


Add to cart

Description

Description

The antibody detects endogenous levels of total DTX1 protein.

Studies in Drosophila have identified this gene as encoding a positive regulator of the Notch-signaling pathway. The human gene encodes a protein of unknown function; however, it may play a role in basic helix-loop-helix transcription factor activity.
Specifications

Specifications

DTX1
Polyclonal
Unconjugated
DTX1
Liquid
DTX1
Synthetic peptide corresponding to a region derived from internal residues of human deltex homolog 1 (Drosophila)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin)
2.1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q86Y01, Q61010
hDx-1
Rabbit
IgG
100 μL
-20°C
Primary
14357, 1840
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit