Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DTYMK Monoclonal Antibody (OTI1E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA503490
Description
Thymidylate kinase (DTYMK) catalyzes the phosphorylation of dTMP to form dTDP in the dTTP synthesis pathway for DNA synthesis. DTYMK is essential for DNA synthesis and is an important intermediate enzyme in the pathway of many pyrimidine analog drugs, including 5-Fluorouracil (5-FU), the chemotherapeutic drug of choice for several solid tumors. The human DTYMK gene was isolated via functional complementation of a Saccharomyces cerevisiae cell cycle mutant, cdc8. DTYMK mRNA levels and enzyme activities are regulated at the level of cell cycle and cell growth pathways.Specifications
DTYMK | |
Monoclonal | |
0.42 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P23919 | |
DTYMK | |
Full length human recombit protein of human DTYMK produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI1E12 | |
Unconjugated | |
DTYMK | |
CDC8, PP3731, TMPK, TYMK | |
Mouse | |
Affinity Chromatography | |
RUO | |
1841 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction