Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTYMK Mouse anti-Human, PE/Atto594, Clone: OTI1G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270604PEATT594

Catalog No. N270604P594


Add to cart

Description

Description

DTYMK Monoclonal antibody specifically detects DTYMK in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DTYMK
OTI1G8
Flow Cytometry
Affinity Purified, Conjugated
DTYMK
Full length recombinant protein of human DTYMK (NP_036277) produced in HEK293T cells.
Immunogen affinity purified
Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary
1841.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
CDC8TMPKTYMKPP3731, deoxythymidylate kinase (thymidylate kinase), dTMP kinase, EC 2.7.4.9, FLJ44192, MGC198617, thymidylate kinase
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit