Learn More
United Scientific Supplies Dual Drive Van de Graaff Generator
SCI_ED
Uniquely offers hand-cranked or motor driven alternative drive modes
$18.00 - $507.00
Specifications
Type Dual Drive
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S27873
SCI_ED
View Documents
United Scientific Supplies
VANDGF1DD
Van De Graaff Generator
Each for $507.00
Add to cart
S27874
SCI_ED
View Documents
United Scientific Supplies
VANDGF1RB
Replacement Belt
Each for $18.00
Add to cart
Description
Description
  • Operates by hand crank or by motor
  • Generates approximately 200,000 volts
  • Produces immediate and lengthy sparks, even in humid conditions
  • Collector sphere crafted from dent- and scratch-resistant polished steel
  • Built-in UV lamp provides additional light
  • Includes accessory kit and activity guide
  • Replacement Belt sold separately
  • Diameter: 28 cm (Collector Sphere)
  • Device: 38 x 23 x 76 cm
Specifications
Specifications
Dual Drive