Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dual-Light™ System Buffer B with Galacton-Plus™ Substrate Non-distribution product as customer accommodation.

Lyophilized luciferin and Galacton-Plus substrate solution are used with the Dual-Light Reporter Gene Assay System. The Dual-Light luminescent reporter gene assay is designed for rapid and sensitive detection of firefly luciferase and E. coli β-galac

Manufacturer:  Invitrogen™ T1053

Catalog No. T1053


Add to cart

Description

Description

Specifications
  • Form: Liquid
  • Format: 96-well plate
  • Substrate: Galacton-Plus, Luciferin
Specifications

Specifications

Buffer B with Galacton-Plus Substrate
96-well plate
Detection of luciferase and β-galactosidase in a single extract aliquot for gene expression assays
200 assays
Enhanced Chemiluminescence
Lyophilized luciferin and Galacton-Plus substrate solution
Luminometer
Store at -20°C
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.