Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Dual Purpose Scoops, White, pk 100

The dual purpose scoop is a unique economical solution that combines the function of a scoop and a weigh boat into one product.

$31.25

Specifications

Color White
Coating Anti-static Polystrene
Disposable Yes
For Use With (Application) Weighing Material
Material PS (Polystyrene)
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
14-127-562
View Documents
Heathrow Scientific™
HS120657
Small Pack of 100 for $31.25
Add to cart
 
14-127-561
View Documents
Heathrow Scientific™
HS120656
Large Pack of 100 for $31.25
Add to cart
 
Description

Description

The weighing process has been streamlined, now just sample or retrieve the material in the scoop and place it directly on the scale. This eliminates the need for a weigh boat or weighing paper and saves valuable space in the lab. When finished the dual solution scoop is disposable.

 

  • Made from anti-static polystyrene that is waterproof and
    reduces the chance of materials sticking to the scoop
  • Ships and stores flat and when ready to use, is easy to
    assemble by inserting the locking tab into the hole
  • Locking tab feature enables a scoop shape to help retain materials
  • Broad tip design keeps the scoop stable while on the scale and
    enables materials to be easily scooped and dispensed
  • Scoop can be used flat as an alternative to weighing paper
  • Easily assemble by pulling the arrow tab through the hole

 

Specifications

Specifications

White
Yes
PS (Polystyrene)
100
Anti-static Polystrene
Weighing Material
Weighing Scoop
No
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit