Learn More
Fisherbrand™ Dual-Purpose Pipets
SDP
Dual-purpose pipets
 View more versions of this product
Catalog No. 11-382-33K
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Borosilicate glass, permanent amber markings, Tuf-Temp™-treated to reduce chipping
  • Accuracy printed on each pipet
  • Capacity marked below mouthpiece
Order Info
Available as PersonaPet™ personalized pipets
Compliance
Calibrated “To Deliver/Blow Out” (upper graduation) and “To Contain” (lower graduation).
Specifications
Specifications
±0.01 mL
0.13 oz.
Dual Purpose Pipet
Borosilicate Glass
4 mL
Product Suggestions
Product Suggestions