Learn More
Brady™ Duct Markers: RETURN AIR
Differentiate between different air systems. Brady™ Duct Markers are made of B-946 self-sticking vinyl.
Manufacturer:  Brady™ 98496
 View more versions of this product
Catalog No. 19-112-039
Add to cart
Description
Description
  • Color-coded to differentiate between different air systems
  • With detachable arrows which can be applied in any direction
  • Made of Pressure Sensitive Vinyl (B-964)
Specifications
Specifications
Acrylic, Permanent, Cold temperature
RETURN AIR
W x H: 20.5 x 4 in.
Vinyl (B-946)
White, Blue
Abrasion and chemical resistant
Die-cut on cards
Duct Marker