Learn More
Mystaire™ Ductless Forensic Workstation Main Filter
For use with the Mystaire™ Ductless Forensic Workstation (FE-36). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
$665.50 - $779.00
Specifications
For Use With (Equipment) FE-36
Product Type Filter
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Application) Material Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Material Price Quantity & Availability  
15-338-315
View Documents
Mystaire™
FE36HF
Particulate Air HEPA
Each for $665.50
Add to cart
15-338-314
View Documents
Mystaire™
FE36CF
Hydrocarbons Gas Phase Carbon
Each for $779.00
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR)
Specifications
Specifications
FE-36
Filter