Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dulcitol, 99%

Click to view available options
Quantity:
250g
500g
608-66-2
C6H14O6
182.172
MFCD00064288
FBPFZTCFMRRESA-GUCUJZIJSA-N
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
11850
CHEBI:16813
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O
608-66-2
C6H14O6
182.172
MFCD00064288
FBPFZTCFMRRESA-GUCUJZIJSA-N
dulcitol, galactitol, dulcite, d-galactitol, dulcose, euonymit, melampyrin, melampyrit, d-dulcitol, 2r,3s,4r,5s-hexane-1,2,3,4,5,6-hexaol
11850
CHEBI:16813
(2R,3S,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O

Useful reagent in bacteriology

Useful reagent in bacteriology

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 1.47
Color White
Melting Point 187°C to 191°C
Boiling Point 275°C to 280°C (1mmHg)
Beilstein 1721903
Merck Index 14,4332
Quantity 250g
Sensitivity Hygroscopic
Formula Weight 182.17
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Dulcitol
Show More Show Less

EINECSNumber : 210-165-2

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only