Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DULLARD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548455
Description
Predicted to react with bovine, mouse and rat based on sequence homology.
Serine/threonine protein phosphatase forming with CNEP1R1 an active phosphatase complex that dephosphorylates and may activate LPIN1 and LPIN2. LPIN1 and LPIN2 are phosphatidate phosphatases that catalyze the conversion of phosphatidic acid to diacylglycerol and control the metabolism of fatty acids at different levels. May indirectly modulate the lipid composition of nuclear and/or endoplasmic reticulum membranes and be required for proper nuclear membrane morphology and/or dynamics. May also indirectly regulate the production of lipid droplets and triacylglycerol. May antagonize BMP signaling.
Specifications
DULLARD | |
Polyclonal | |
Unconjugated | |
CTDNEP1 | |
2610507E10Rik; CTD nuclear envelope phosphatase 1; CTDNEP1; C-terminal domain nuclear envelope phosphatase 1; Dullard; dullard homolog; HSA011916; NET56; serine/threonine-protein phosphatase dullard | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23399 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
O95476 | |
CTDNEP1 | |
KLH conjugated synthetic peptide between 131-160 amino acids from the Central region of human DULLARD. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction