Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DUOXA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548575

Catalog No. PA548575

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Required for the maturation and the transport from the endoplasmic reticulum to the plasma membrane of functional DUOX2. May play a role in thyroid hormone synthesis.
Specifications

Specifications

DUOXA2
Polyclonal
PBS with 0.09% sodium azide
Q1HG44
Dual oxidase maturation factor 2, Dual oxidase activator 2, DUOXA2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
405753
Western Blot
Unconjugated
DUOXA2
Liquid
DUOXA2
KLH conjugated synthetic peptide between 286-314 amino acids from the C-terminal region of human DUOXA2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit