Alfa Aesar™ Duplex 2209, 40 mesh woven from 0.2mm (0.008mm) dia wire

Manufacturer: Alfa Aesar™ 46504GT

Quantity 150 x 150mm

 View more versions of this product

Catalog No. AA46504GT

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Duplex 2209, 40 mesh woven from 0.2mm (0.008mm) dia. wire

    Specifications

    Quantity 150 x 150mm
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Wire Cloth
    Health Hazard 2 GHS H Statement
    H351-H373-H317
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    Mesh Size 40 Mesh
    Odor Odorless
    Health Hazard 1 Warning
    Chemical Name or Material Duplex 2209, 40 mesh woven from 0.2mm (0.008mm) dia. wire

    GHS H Statement
    H351-H373-H317
    Suspected of causing cancer.
    May cause damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit