Learn More
Axxora LLCSupplier Diversity Partner DUPLEX-SPECIFIC NUCLEASE LYOPH DFS Item
Manufacturer:  Axxora LLCSupplier Diversity Partner EVNEA003KI01
Duplex-Specific Nuclease Kit (10 Units). Duplex-Specific Nuclease (DSN) enzyme purified from Kamchatka crab hepatopancreas. DSN displays a strong preference for cleaving double-stranded DNA and DNA in DNA-RNA hybrid duplexes, compared to single-stranded DNA. Moreover, the cleavage rate of short, perfectly matched DNA duplexes by this enzyme is essentially higher than that for non-perfectly matched duplexes of the same length.
Catalog No. NC0165500