Learn More
DuPont™ DuPont ™ Fully Encapsulated Level A Tychem ™ TK ™ Suits, NFPA 1991
Excellent chemical holdout for use in hazmat situations
Manufacturer:  DuPont™ TK600TLYXL000100
 View more versions of this product
Catalog No. 19-121-753
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Made from a high-strength nonwoven with nonhalogenated films on either side.
Inner Suit
  • Double-taped seams
  • Three-layer EX faceshield (PVC 40mil/PTFE 5mil/PVC 20mil)
  • Gastight zipper closure
  • Double-storm flap
  • Attached three-layer gloves (Barrier™/neoprene/Kevlar™ knit) and sock boots
Outer Suit
  • Aluminized fiberglass with sewn NOMEX™ seams
  • EX faceshield (Lexan™ 20mil)
  • Hook-and-loop closure
  • Storm flap
  • Hemmed wrists
Specifications
Specifications
NFPA 1991
Silver
5.9 to 6.3 ft.
Industrial, HazMat and domestic preparedness applications
74.29cm
78.105cm
27mil
Expanded faceshield, 4 exhaust valves, internal adjustment waist belt, attached internal North SSG - Silver Shield™/4H™/external Neoprene size 10 gloves, separate Kevlar™ knit over gloves, socks with outer boot flaps, soles, separate Aluminize; Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant
118.74 to 127.65cm
Taped
X-Large
1.79 to 1.92m
29.25 in.
30.75 in.
Front entry
Zipper front
Tychem™ TK
46.75 to 50.25 in.