DuPont ™ General Environment Series 181 Coveralls

Air permeability helps keep wearer comfortable

Manufacturer: Dupont Personal Protection GE181SWH2X00250B

 View all options for this product

Catalog No. 17-685-308


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Collar Type Lay down
    Closure Type Zipper
    Ankle Style Elastic
    Cuff Style Elastic
    Material Polypropylene SMS
    Seam Type Serged
    Color White
    Size 2X-Large
    Size (English) Chest 28.75 in.
    Disposable Yes
    Size (Metric) Chest 73.02cm
    For Use With (Application) General environment
    Size (English) Inseam 30.5 in.
    Size (English) Waist 56 in.
    Size (Metric) Inseam 77.47cm
    Size (Metric) Waist 142.24cm

    Made from polypropylene SMS fabric that resists nonhazardous dry particle penetration.

    • Antistatic treated
    • Serged seams
    • Laydown collar
    • Set sleeves
    • Elastic wrists and ankles
    • Zipper closure
    • White or blue

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit