Learn More
DuPont™ General Environment Series 219 Lab Coats
Antistatic treated
Specifications
Collar Type Knit
Closure Type Snaps
Cuff Style Knit
Material Polypropylene, SMS
No. of Pockets 3
View More Specs
Products 2
Catalog Number Mfr. No. Color Size Length (English) Sleeve Size (English) Chest Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Length (English) Sleeve Size (English) Chest Price Quantity & Availability  
17-685-328
View Documents
DuPont™
GE219SWHXL00300B
White X-Large 40.75 in. 38 to 40 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
17-685-326
View Documents
DuPont™
GE219SWHMD00300B
White Medium 38.75 in. 32 to 36 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Made from antistatic-treated polypropylene SMS fabric that resists nonhazardous dry particle penetration.
  • Serged seams
  • Knit collar and cuffs
  • Set sleeves
  • Six front snap entry with adjustable top snap
  • Three pockets (one left chest, two lower front)
  • Nonsterile
  • White or blue
Order Info
Bulk packaged in double transparent poly liners
Specifications
Specifications
Knit
Knit
3
General environment
Non-sterile
Snaps
Polypropylene, SMS
Yes
Serged