Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed

Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.

Manufacturer:  DuPont™ IC182BWHXL00250C

 View more versions of this product

Catalog No. 19-088-1121D


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Zipper
40 to 43 in.
Elastic,Tunnelized
White
5.11 to 6.3 ft.
Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
99.06cm
86.36cm
Tyvek™ IsoClean™ Flashspun Polyethylene
101.6 to 109.22cm
Bound
X-Large
155.75 to 192.04cm
39 in.
34 in.
5.4mil
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit