DuPont™ Tyvek™ IsoClean™ Series 182B Coveralls, Clean-Processed

Tyvek IsoClean delivers an ideal balance of protection, durability and comfort. Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.

Manufacturer: DuPont™ IC182BWH2X00250C

 View more versions of this product

Catalog No. 19-088-1121E

  Check Availability
Add to Cart

    Specifications

    Material Tyvek™ IsoClean™ Flashspun Polyethylene
    Seam Type Bound
    Color White
    Height (Metric) 185.92 to 198.12cm
    For Use With (Application) Premium protection for controlled environments and excellent inherent barrier to dry particles, microorganisms and non-hazardous liquids
    Length (Metric) Sleeve 101.6cm
    Closure Type Zipper
    Size (English) Inseam 35 in.
    Size (Metric) Inseam 88.9cm
    Thickness 5.4mil
    Size 2X-Large
    Size (English) Chest 43 to 46 in.
    Size (Metric) Chest 109.2 to 116.84cm
    Height (English) 6.1 to 6.5 ft.
    Length (English) Sleeve 40 in.
    Cuff Style Tunnelized elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit