Learn More
DuPont Tyvek IsoClean 270 Series Frocks
No pockets to trap contamination.
Manufacturer: DuPont IC270BWHMD00300C
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Made of Tyvek, a spunbonded olefin fabric that is lightweight and flexible, yet provides superior contamination control and particulate filtration, unsurpassed liquid and particulate shielding and two-way contamination protection. Garments are constructed with the smooth side out to reduce particulate entrapment and bound seams reduce lint. Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags. ISO Class 5 (formerly Class 100) compatible. Constructed to hold up and keep you comfortable. Generous sizing permits freedom of movement. Plastic snaps won't rust or flake during processing. Available in bulk (no special cleaning), clean (ISO Class 4 [formerly Class 10] cleanroom processed), sterile (gamma irradiated), or clean and sterile. Every lot is 100% traceable.
- Excellent barrier to dry particulates, nonhazardous liquids, and microorganisms
- Bound neck with adjustable snaps Five snap front Tunneled elastic wrist Made from DuPont Tyvek brand protective material Unique, patented flashspinning process creates an excellent barrier to dry particles, microorganisms and nonhazardous liquids Comfortable, lightweight and durable Full traceability on all sterilized apparel Recyclable
Specifications
35.5 in. | |
32 to 36 in. | |
White | |
101.6 cm | |
Bound | |
Clean-processed |
90.17 cm | |
81 to 91 cm | |
40 in. | |
Tyvek™ | |
Medium |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.