Learn More
DuPont™ NexGen™ Protective Coveralls: With Collar
Description
Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.
- Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
- Proprietary bilaminate composite fabric provides superior liquid penetration1
- Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
- Low linting for use in critical applications
- Long torso with polyester front zipper measures 26 in. long (66cm)
- White
1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).
Specifications
Size | Large |
Material | Fabric |
Color | White |
Disposable | Single-use |
Ankle Style | Open |
Cuff Style | Open |
Neck Style | Collared |
Closure Type | Front Zipper |
Seam Type | Serged |
Collar Type | Lay down |
Show More |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.