Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ NexGen™ Protective Coveralls: With Collar

Catalog Number 190882140
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Affordable high-barrier protection

Lightweight coveralls (1.8 oz. per sq. yd.) provide excellent barrier protection against nontoxic particle, liquid and spray hazards. True aerosol- and liquid-proof microporous barrier demonstrates >99% holdout of particles down to 0.35μm.

  • Breathable, cloth-like material treated to provide antistatic properties for use in controlled environments
  • Proprietary bilaminate composite fabric provides superior liquid penetration1
  • Compared favorably to leading fabric in physical property tests covering resistance to ripping and tearing2
  • Low linting for use in critical applications
  • Long torso with polyester front zipper measures 26 in. long (66cm)
  • White

1 Measured per ASTM™ F 903 in relation to the following substances: household bleach, diazinon (4 oz./gal.), hydraulic jack oil, machine oil, malathion (50%), motor oil (40W), sodium hydroxide and isocyanate-based paint hardener. Tests were performed by independent laboratories on material samples under laboratory conditions, not with actual garments or in actual onsite conditions.
2 ASTM D 751 (Grab Tensile Strength), ASTM D 1117 (Trapezoid Tear), ASTM D 3787 (Ball Burst).

TRUSTED_SUSTAINABILITY
Size X-Large
Material Fabric
Color White
Disposable Single-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Collar Type Lay down
Certifications/Compliance ASTM D751, D1117, D5034, D257, AATCC 127
Description Microporous Film, Laminated to a Spundbonded Polypropylene Substrate
Size (English) Chest 40.75 to 44.25 in.
Size (English) Inseam 28.25 in.
Size (Metric) Chest 103.5 to 112.39 cm
Size (Metric) Inseam 71.75 cm
Thickness 10 mil
For Use With (Application) Automotive refinishing, waste cleanup and remediation
Length (English) Sleeve 35 in.
Length (Metric) Sleeve 88.9 cm
Height (English) 5.8 to 6.2 ft.
Height (Metric) 1.76 to 1.88 m
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit