DuPont™ ProShield™ Basic Coveralls with Attached Hood and Elastic Ankles

Provides comfortable, quality coverall to workers for nonhazardous dry particle applications

Manufacturer: Dupont Personal Protection PB120SBU7X002500

 View more versions of this product

Catalog No. 19-162-647

  Check Availability
Add to Cart

    Specifications

    Material SMS
    Seam Type Serged
    Color Blue
    Size 6X-Large
    Height (English) Not Applicable
    Height (Metric) Not Applicable
    For Use With (Application) General maintenance, janitorial/cleaning and other dirty work
    Includes Hood, elastic wrist and ankle
    Disposable Yes
    Closure Type Zipper Front with Storm Flap

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit