Learn More
DuPont™ ProShield™ 10 Coveralls
Catalog Number 19162554
Click to view available options
Size:
7X-Large
Color:
White
This item is not returnable. View return policy
This item is not returnable. View return policy
Provides comfortable, quality coverall to workers for nonhazardous dry particle applications
DuPont™ ProShield™ 10, spunbond meltblown spunbond (SMS) garments, are made to high quality standards and have been optimized for comfort, softness and breathability while offering the cost advantages of disposable garments 
DuPont™ ProShield™ 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Applications include: general maintenance, janitorial/cleaning and other dirty work.
  • Attached hood with elastic around face opening
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Front zipper closure with storm flap