Learn More
DuPont™ ProShield™ 10 Coveralls with Elastic Wrists and Ankles
Provide comfortable, quality coverall to workers for nonhazardous dry particle applications
Supplier: DuPont™ PB127SBU6X002500
Description
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles. Applications include: general maintenance, janitorial/cleaning and other dirty work.
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles in a choice of white, blue and gray. Applications include: general maintenance, janitorial/cleaning and other dirty work.Compliance
Meets ASTM™ D1117 standards.
Specifications
6X-Large | |
Blue | |
Elastic | |
Serged | |
General maintenance, janitorial/cleaning and other dirty work |
SMS | |
Yes | |
Front zipper with storm flap | |
Coveralls, elastic wrists |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.