Learn More
DuPont™ ProShield™ 10 Coveralls with Elastic Wrists and Ankles
Catalog Number 19162646
This item is not returnable. View return policy
This item is not returnable. View return policy
Provide comfortable, quality coverall to workers for nonhazardous dry particle applications
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles. Applications include: general maintenance, janitorial/cleaning and other dirty work.
DuPont ProShield 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles in a choice of white, blue and gray. Applications include: general maintenance, janitorial/cleaning and other dirty work.
Compliance
Meets ASTM™ D1117 standards.