DuPont™ ProShield™ Basic Coveralls with Elastic Wrists and Ankles

Provide comfortable, quality coverall to workers for nonhazardous dry particle applications

Manufacturer: Dupont Personal Protection PB125SWHXL002500

 View more versions of this product

Catalog No. 19-162-530

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material SMS
    Includes Hood, elastic wrist, attached skid-resistant boots
    Seam Type Serged
    Color White
    Size X-Large
    Disposable Yes
    Height (English) 5.8 to 6.2 ft.
    Height (Metric) 1.76 to 1.89m
    For Use With (Application) General maintenance, janitorial/cleaning and other dirty work
    Length (Metric) Sleeve 87.6cm
    Closure Type Zipper Front with Storm Flap
    Size (English) Inseam 27.75 in.
    Size (Metric) Inseam 70.48cm
    Size (English) Chest 40.75 to 44.25 in.
    Size (Metric) Chest 103.6 to 112.4cm
    Length (English) Sleeve 34.5 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit