Learn More
DuPont™ ProShield™ Basic Coveralls with Open Wrists and Ankles
Catalog Number 19162510
This item is not returnable. View return policy
This item is not returnable. View return policy
Provide comfortable, quality coverall to workers for nonhazardous dry particle applications
  • Soft, breathable and lightweight with comfort fit design
  • Spunbond Meltblown Spunbond (SMS)
  • Disposable
  • Laydown collar
  • Open wrists, ankles
  • Front zipper closure
  • Serged seams
  • Available in blue or white