Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ Basic Coveralls with Open Wrists and Ankles

Provide comfortable, quality coverall to workers for nonhazardous dry particle applications

Manufacturer:  Dupont Personal Protection PB120SBULG002500

 View more versions of this product

Catalog No. 19-162-508


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Zipper Front with Storm Flap
37.25 to 40.75 in.
Open
Open
Blue
5.5 to 5.9 ft.
General maintenance, janitorial/cleaning and other dirty work
83.18cm
SMS
94.6 to 103.5cm
Yes
Serged
Large
1.67 to 1.8m
32.75 in.
69.85cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit