DuPont™ ProShield™ Basic Coveralls with Elastic Wrists and Ankles PROMO

Provide comfortable, quality coverall to workers for nonhazardous dry particle application

Manufacturer: Dupont Personal Protection PB125SBUXL002500PMO

 View more versions of this product

Catalog No. 19-162-537PMO

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Material SMS
    Seam Type Serged
    Color Blue
    Size X-Large
    Disposable Yes
    For Use With (Application) General maintenance, janitorial/cleaning and other dirty work
    Closure Type Zipper Front with Storm Flap

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit