DuPont™ Proshield™ Basic Disposable Lab Coats
Designed for workers in a range of industries for nonhazardous dry particle applications
Manufacturer: Dupont Personal Protection PB219SBU3X003000
 View more versions of this product
Catalog No. 19-162-624
  Check Availability
Add to Cart
    Specifications
    Product Type Disposable lab coat
    Material SMS (Spunbound Melt-Blown Synthetic)
    Size 3X-Large
    Disposable Yes
    Collar Type Knit
    Latex Free Yes
    Seam Type Serged
    Closure Type Snap
    Color Blue
    Cuff Style Knit
  • Description & Specifications
Related Products
Product Content Feedback