Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont Proshield 10 Basic Disposable Lab Coats
Designed for workers in a range of industries for nonhazardous dry particle applications
Specifications
Collar Type | Knit |
---|---|
Disposable | Yes |
Latex Free | Yes |
Seam Type | Serged |
Closure Type | Snaps |
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Size | Price | Quantity & Availability | ||||
19-162-621
|
DuPont
PB219SBULG003000 |
Blue | Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-620
|
DuPont
PB219SBUMD003000 |
Blue | Medium |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-619
|
DuPont
PB219SBUSM003000 |
Blue | Small |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-622
|
DuPont
PB219SBUXL003000 |
Blue | X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-624
|
DuPont
PB219SBU3X003000 |
Blue | 3X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-623
|
DuPont
PB219SBU2X003000 |
Blue | 2X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-625
|
DuPont
PB219SBU4X003000 |
Blue | 4X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-626
|
DuPont
PB219SWHSM003000 |
White | Small |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-627
|
DuPont
PB219SWHMD003000 |
White | Medium |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-628
|
DuPont
PB219SWHLG003000 |
White | Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-629
|
DuPont
PB219SWHXL003000 |
White | X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-630
|
DuPont
PB219SWH2X003000 |
White | 2X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-631
|
DuPont
PB219SWH3X003000 |
White | 3X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-162-632
|
DuPont
PB219SWH4X003000 |
White | 4X-Large |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Disposable lab coats that are optimized for comfort, softness and breathability
- Spunbond meltblown spunbond (SMS) material
- Three pockets (one left chest pencil, two lower front)
- Snap closure (six plus one adjustable)
- Serged seams
- Set sleeves
- Knit collar
- Knit cuffs
Compliance
Meets ASTM D1777 standards.
Specifications
Knit | |
Yes | |
Snaps | |
SMS |
Yes | |
Serged | |
Knit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title