DuPont™ ProShield™ Basic Sngle Use Coveralls with Elastic Wrists and Ankle PROMO

Provide comfortable, single use coverall protection to workers for nonhazardous dry particle applications

Manufacturer: Dupont Personal Protection PB125SWH6X002500PMO

 View more versions of this product

Catalog No. 19-162-643PMO

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Material SMS
    Size 6X-Large
    Seam Type Serged
    Size (English) Chest 59.25 - 62.75 in.
    For Use With (Application) General maintenance and hospital visitation
    Disposable Yes
    Collar Type Laydown
    Length (English) Sleeve 41 in.
    Color White
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit