Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProShield™ NexGen™ Sleeves
Click to view available options
Sleeves; 18 in. (46cm) long
Description
DuPont ProShield NexGen is liquid resistant and provides a true aerosol barrier, with >99% holdout of particles down to 0.35μm. Proprietary microporous technology gives sleeves effective protective properties at an economical price. Perfect for applications involving nontoxic liquid or spray hazards.
Sleeves
- Elastic at both ends
- Length: 18 in. (46cm)
- White
Specifications
Product Type | General Purpose Sleeve |
Material | Microporous Film Laminate |
Disposable | Single-use |
Seam Type | Serged |
Thickness | 10 mil |
Packaging | 100 Pairs/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction