Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ ProShield™ NexGen™ Apron

Catalog Number 19048402a
Click to view available options
Apron; L x W: 36 x 28 in. (69 x 71cm)
This item is not returnable. View return policy
This item is not returnable. View return policy

Maximum protection at affordable cost

DuPont ProShield NexGen is liquid resistant and provides a true aerosol barrier, with >99% holdout of particles down to 0.35μm. Proprietary microporous technology gives aprons effective protective properties at an economical price. Perfect for applications involving nontoxic liquid or spray hazards.

Apron

  • Bib-style apron provides coverage of front and sides
  • With bound neck and ties
  • Length: 36 in. (91cm)
  • White
TRUSTED_SUSTAINABILITY
Material Tyvek
Color White
Closure Type Ties
Includes Case of 100
Length (English) 36 in.
Length (Metric) 69 cm
Seam Type Bound
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit