DuPont ™ ProShield ™ NexGen ™ Shoe Covers

Manufacturer: Dupont Personal Protection NG451SWHSM020000

 View more versions of this product

Catalog No. 19-715-310

  Check Availability
Add to Cart

    Specifications

    Sole Type Non-skid
    Material ProShield™ NexGen™, Microporous, Spundbonded Polypropylene
    Length (English) 9.75 in.
    Color White
    Size Small
    Product Type Shoe Covers
    For Use With (Application) Ideal for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications and automotive refinishing, waste cleanup, remediation
    Height (English) 5 in.
    Closure Type Elastic
    Height (Metric) 12.7cm
    Length (Metric) 24.76cm
    Seam Type Serged
    Thickness 10mil

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit