Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ ProShield™ NexGen™ Shoe Covers
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
ProShield 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications. ProShield 60 garments are made with a microporous film that is laminated to a spundbonded polypropylene substrate. Applications include: automotive refinishing, waste cleanup and remediation.5 in. shoe cover with elastic top PVC soles provides enhanced skid-resistance and durability. One size fits most.
Specifications
Size | Large |
Material | Polypropylene |
Color | White |
Closure Type | Elastic Ankle |
Sole Type | Non-skid |
Model | NG451SWHLG020000 |
Seam Type | Serged |
Thickness | 10 mil |
For Use With (Application) | Ideal for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications and automotive refinishing, waste cleanup, remediation |
Length (English) | 14 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction