DuPont ™ Spunbonded Polypropylene Shoe Covers

Available with or without skid-resistant tread

Manufacturer: Dupont Personal Protection 2852

 View more versions of this product

Catalog No. 19-130-2727

  Check Availability
Add to Cart

    Specifications

    Sole Type Skid-resistant
    Size Universal
    Product Type Shoe Covers
    Material Spunbond Polypropylene, Poly Film
    For Use With (Application) Ideal for the challenges of dry and wet condition
    Latex Free Yes
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit