DuPont ™ Suprel ™ LS Boot Covers
Developed for life science, pharmaceutical, biotechnology, and medical device manufacturing industries
Manufacturer: Dupont Personal Protection SR447SBUXL010000
 View more versions of this product
Catalog No. 19-171-006
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Size X-Large
    Patented Suprel LS bi-component fabric offers breathability and barrier protection combined with soft feel and optimal comfort.
    • High-top style extends 17.5 in. (45cm) up leg
    • Serged elastic at opening with elastic ankle
    • Color: White
    Suprel LS is not flame-resistant. Should not be used around heat, flame, sparks, or in potential flammable or explosive environments.
  • Description & Specifications
Related Products
Product Content Feedback