DuPont ™ Suprel ™ LS Frocks
Suit a wide range of applications requiring comfortable protection
Manufacturer: Dupont Personal Protection SR267SWH2X003000
 View all options for this product
Catalog No. 19-165-192
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Color White
    Size 2X-Large
    Patented Suprel LS bi-component fabric offers breathability and barrier protection combined with soft tactile and optimal comfort. Developed specifically for life science, pharmaceutical, biotechnology, and medical device manufacturing industries. White.
    • Five-snap front-closure
    • Mandarin collar
    • Full sleeves with tunnel elastic wrists
    • No pockets
    • Color: White
  • Description & Specifications
Related Products
Product Content Feedback